missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoropyridine-3-carboxaldehyde, 95%

Catalog No. AC449850010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449850010 1 g Glass bottle
AC449850050 5 g Glass bottle
AC449850250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449850010 Supplier Thermo Scientific Chemicals Supplier No. 449850010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 677728-92-6 | C6H4FNO | 125.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 677728-92-6
Molecular Formula C6H4FNO
Molecular Weight (g/mol) 125.1
InChI Key PZPNGWWKCSJKOS-UHFFFAOYSA-N
Synonym 2-fluoropyridine-5-carbaldehyde, 6-fluoronicotinaldehyde, 2-fluoro-5-formylpyridine, 2-fluoropyridine-5-carboxaldehyde, 6-fluoro-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 6-fluoro, 6-fluoropyridine-3-carboxaldehyde, pubchem5150, zlchem 1162, acmc-1b2mv
PubChem CID 16414246
IUPAC Name 6-fluoropyridine-3-carbaldehyde
SMILES C1=CC(=NC=C1C=O)F

Specifications

Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 125.1
Percent Purity 95%
Chemical Name or Material 6-Fluoropyridine-3-carboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only