missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoronicotinic Acid 98.0+%, TCI America™

Catalog No. F0695200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0695200MG 200 mg
F06951G 1 g
F06955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F0695200MG Supplier TCI America Supplier No. F0695200MG

Chemical Identifiers

CAS 403-45-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD01859863
InChI Key UJDLCTNVHJEBDG-UHFFFAOYSA-N
Synonym 6-fluoronicotinic acid, 2-fluoropyridine-5-carboxylic acid, 6-fluoro-3-pyridinecarboxylic acid, 2-fluoro-5-carboxypyridine, 2-fluoro-5-pyridinecarboxylic acid, 3-carboxy-6-fluoropyridine, 3-pyridinecarboxylic acid, 6-fluoro, 6-fluoro-nicotinic acid, 6-fluoronicotinc acid, 2-fluoro-5-pyridine carboxylic acid
PubChem CID 242819
IUPAC Name 6-fluoropyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(F)N=C1

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 200 mg
Formula Weight 141.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoronicotinic Acid