missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoronicotinic acid, 97%

Catalog No. p-7036770
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55147MD 250 mg
AAH5514703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55147MD Supplier Thermo Scientific Chemicals Supplier No. H55147MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alfa Aesar 6-Fluoronicotinic acid, 97%, Quantity: 1g, Melting Point: 144 deg.C to 148 deg.C, Molecular Weight: 141.101, Percent Purity: 97%, CAS: 403-45-2, Formula Weight: 141.1, Health Hazard 1: H315-H319-H335 | CAS: 403-45-2 | C6H4FNO2 | 141.10 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.

Chemical Identifiers

CAS 403-45-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD01859863
InChI Key UJDLCTNVHJEBDG-UHFFFAOYSA-N
Synonym 6-fluoronicotinic acid, 2-fluoropyridine-5-carboxylic acid, 6-fluoro-3-pyridinecarboxylic acid, 2-fluoro-5-carboxypyridine, 2-fluoro-5-pyridinecarboxylic acid, 3-carboxy-6-fluoropyridine, 3-pyridinecarboxylic acid, 6-fluoro, 6-fluoro-nicotinic acid, 6-fluoronicotinc acid, 2-fluoro-5-pyridine carboxylic acid
PubChem CID 242819
IUPAC Name 6-fluoropyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(F)N=C1

Specifications

Melting Point 144°C to 148°C
Quantity 250 mg
Formula Weight 141.1
Percent Purity 97%
Chemical Name or Material 6-Fluoronicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only