missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluoroindole 98.0+%, TCI America™
$70.84 - $212.51
Chemical Identifiers
| CAS | 399-51-9 |
|---|---|
| Molecular Formula | C8H6FN |
| Molecular Weight (g/mol) | 135.14 |
| MDL Number | MFCD00056933 |
| InChI Key | YYFFEPUCAKVRJX-UHFFFAOYSA-N |
| Synonym | 6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g |
| PubChem CID | 351278 |
| IUPAC Name | 6-fluoro-1H-indole |
| SMILES | FC1=CC=C2C=CNC2=C1 |
Chemical Identifiers
| 399-51-9 | |
| 135.14 | |
| YYFFEPUCAKVRJX-UHFFFAOYSA-N | |
| 351278 | |
| FC1=CC=C2C=CNC2=C1 |
| C8H6FN | |
| MFCD00056933 | |
| 6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g | |
| 6-fluoro-1H-indole |
Specifications
| 399-51-9 | |
| C8H6FN | |
| 1 g | |
| YYFFEPUCAKVRJX-UHFFFAOYSA-N | |
| 6-fluoro-1H-indole | |
| 351278 | |
| ≥98.0% (GC) | |
| 6-Fluoroindole |
| 76°C | |
| MFCD00056933 | |
| 6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g | |
| FC1=CC=C2C=CNC2=C1 | |
| 135.14 | |
| 135.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No