missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroindole-3-carboxaldehyde, 98%

Catalog No. AAH6166506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6166506 5 g
AAH6166503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6166506 Supplier Thermo Scientific Chemicals Supplier No. H6166506
Only null left

CAS: 2795-41-7 | C9H6FNO | 163.15 g/mol

6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in methanol.

Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.

Chemical Identifiers

CAS 2795-41-7
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00069703
InChI Key CWCYUOSLRVAKQZ-UHFFFAOYSA-N
Synonym 6-fluoroindole-3-carboxaldehyde, 6-fluoroindole-3-carbaldehyde, 6-fluoroindole-3-aldehyde, 6-fluoro-1h-indole-3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 6-fluoro, 1h-6-fluoroindole-3-carboxaldehyde, pubchem7269, ksc495g4p, 6-fluoro-3-formyl-1h-indole, 6-fluorooindole-3-carboxaldehyde
PubChem CID 262903
IUPAC Name 6-fluoro-1H-indole-3-carbaldehyde
SMILES FC1=CC=C2C(NC=C2C=O)=C1

Specifications

Color Yellow
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 163.15
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 6-Fluoroindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only