missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluorochromone-3-carboxaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 157°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 192.15 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Fluorochromone-3-carboxaldehyde |
Safety and Handling
| TSCA | No |