missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-4H-1,3-benzodioxine-8-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC01901EA

 View more versions of this product

Catalog No. CC01901EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

6-Fluoro-4H-1, 3-benzodioxine-8-carboxylic acid
96.5
97%
MFCD02681981
6-fluoro-4h-benzo 1,3 dioxine-8-carboxylic acid, 6-fluoro-2,4-dihydro-1,3-benzodioxine-8-carboxylic acid, 6-fluoro-4h-benzo d 1,3 dioxine-8-carboxylic acid, 6-fluoro-4h-1,3-benzodioxin-8-carboxylic acid, 8-carboxy-6-fluoro-4h-1,3-benzodioxine, 6-fluoro-1,3-benzodioxene-8-carboxylic acid, 4h-1,3-benzodioxin-8-carboxylicacid, 6-fluoro, 6-fluoranyl-4h-1,3-benzodioxine-8-carboxylic acid, 6-fluoro-2h,4h-benzo e 1,3-dioxane-8-carboxylic acid, fbg
C1C2=C(C(=CC(=C2)F)C(=O)O)OCO1
198.149
198.15
321309-28-8
100.0
C9H7FO4
10g
HWBALMSPYAUMMB-UHFFFAOYSA-N
6-fluoro-4H-1,3-benzodioxine-8-carboxylic acid
2779901
97%

Chemical Identifiers

321309-28-8
198.149
HWBALMSPYAUMMB-UHFFFAOYSA-N
2779901
C1C2=C(C(=CC(=C2)F)C(=O)O)OCO1
C9H7FO4
MFCD02681981
6-fluoro-4h-benzo 1,3 dioxine-8-carboxylic acid, 6-fluoro-2,4-dihydro-1,3-benzodioxine-8-carboxylic acid, 6-fluoro-4h-benzo d 1,3 dioxine-8-carboxylic acid, 6-fluoro-4h-1,3-benzodioxin-8-carboxylic acid, 8-carboxy-6-fluoro-4h-1,3-benzodioxine, 6-fluoro-1,3-benzodioxene-8-carboxylic acid, 4h-1,3-benzodioxin-8-carboxylicacid, 6-fluoro, 6-fluoranyl-4h-1,3-benzodioxine-8-carboxylic acid, 6-fluoro-2h,4h-benzo e 1,3-dioxane-8-carboxylic acid, fbg
6-fluoro-4H-1,3-benzodioxine-8-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions