missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(6-Fluoro-4H-1,3-benzodioxin-8-yl)methanol, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC01909DA
Specifications
| (6-Fluoro-4H-1, 3-benzodioxin-8-yl)methanol | |
| 89.5 | |
| 90% | |
| C9H9FO3 | |
| 1 g | |
| YLADDVZIEBXWQU-UHFFFAOYSA-N | |
| (6-fluoro-4H-1,3-benzodioxin-8-yl)methanol | |
| 2779909 | |
| 90% |
| 306934-89-4 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD02677744 | |
| 6-fluoro-4h-1,3-benzodioxin-8-yl methanol, 6-fluoro-4h-benzo d 1,3 dioxin-8-yl methanol, 6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl methanol, 4h-1,3-benzodioxin-8-methanol,6-fluoro, 6-fluoranyl-4h-1,3-benzodioxin-8-yl methanol, 6-fluoro-8-hydroxymethyl-4h-1,3-benzodioxine, 6-fluoro-2h,4h-benzo e 1,3-dioxan-8-yl methan-1-ol | |
| C1C2=CC(=CC(=C2OCO1)CO)F | |
| 184.166 | |
| 184.17 |
Chemical Identifiers
| 306934-89-4 | |
| 184.166 | |
| YLADDVZIEBXWQU-UHFFFAOYSA-N | |
| 2779909 | |
| C1C2=CC(=CC(=C2OCO1)CO)F |
| C9H9FO3 | |
| MFCD02677744 | |
| 6-fluoro-4h-1,3-benzodioxin-8-yl methanol, 6-fluoro-4h-benzo d 1,3 dioxin-8-yl methanol, 6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl methanol, 4h-1,3-benzodioxin-8-methanol,6-fluoro, 6-fluoranyl-4h-1,3-benzodioxin-8-yl methanol, 6-fluoro-8-hydroxymethyl-4h-1,3-benzodioxine, 6-fluoro-2h,4h-benzo e 1,3-dioxan-8-yl methan-1-ol | |
| (6-fluoro-4H-1,3-benzodioxin-8-yl)methanol |