missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole Hydrochloride 98.0+%, TCI America™

Catalog No. F09445G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09445G 5 g
F09441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09445G Supplier TCI America Supplier No. F09445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84163-13-3
Molecular Formula C12H14ClFN2O
Molecular Weight (g/mol) 256.705
MDL Number MFCD06658529
InChI Key CWPSRUREOSBKBQ-UHFFFAOYSA-N
PubChem CID 11334359
IUPAC Name 6-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride
SMILES C1CNCCC1C2=NOC3=C2C=CC(=C3)F.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 256.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole Hydrochloride