missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2-methyl-4-chromanone 97.0+%, TCI America™

Catalog No. F02715G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02715G Supplier TCI America Supplier No. F02715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88754-96-5
Molecular Formula C10H9FO2
Molecular Weight (g/mol) 180.18
MDL Number MFCD00059072
InChI Key RPAIBTVEPAACRP-UHFFFAOYNA-N
PubChem CID 2737390
IUPAC Name 6-fluoro-2-methyl-3,4-dihydro-2H-1-benzopyran-4-one
SMILES CC1CC(=O)C2=C(O1)C=CC(F)=C2

Specifications

Melting Point 71°C
Quantity 5 g
Formula Weight 180.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-2-methyl-4-chromanone