missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2-methyl-3-nitropyridine 97.0+%, TCI America™

Catalog No. F10671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10671G 1 g
F10675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10671G Supplier TCI America Supplier No. F10671G
Only null left

Chemical Identifiers

CAS 18605-16-8
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD00234311
InChI Key JVLRFRZVCOWBDY-UHFFFAOYSA-N
PubChem CID 12408185
IUPAC Name 6-fluoro-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(=N1)F)[N+](=O)[O-]

Specifications

Boiling Point 85°C
Quantity 1 g
Synonym 2-fluoro-5-nitro-6-picoline, 2-fluoro-6-methyl-5-nitropyridine, 2-fluoro-5-nitro-6-methylpyridine, pyridine, 6-fluoro-2-methyl-3-nitro, pubchem6327, acmc-209enj, 6-fluoro-3-nitro-2-picoline, 2-fluoro-5-nitro-6-methylpyridin, 2-fluoro-5-nitro-6-methyl pyridine, pyridine,6-fluoro-2-methyl-3-nitro
Formula Weight 156.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Fluoro-2-methyl-3-nitropyridine