missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-1-indanone 98.0+%, TCI America™

Catalog No. f06685g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06685G 5 g
F06681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06685G Supplier TCI America Supplier No. F06685G
Only null left

Chemical Identifiers

CAS 1481-32-9
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.15
MDL Number MFCD01318147
InChI Key LVUUCFIQQHEFEJ-UHFFFAOYSA-N
Synonym 6-fluoro-1-indanone, 6-fluoro-2,3-dihydro-1h-inden-1-one, 6-fluoro-indan-1-one, 6-fluoroindan-1-one, 6-fluroro-1-indanone, 1h-inden-1-one, 6-fluoro-2,3-dihydro, 6-fluoroindanone, 6fluoro-1-indanone, pubchem8856, acmc-1bwfy
PubChem CID 1519464
IUPAC Name 6-fluoro-2,3-dihydro-1H-inden-1-one
SMILES FC1=CC2=C(CCC2=O)C=C1

Specifications

Melting Point 60°C
Boiling Point 60°C
Quantity 5 g
Formula Weight 150.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-1-indanone