missing translation for 'onlineSavingsMsg'
Learn More

6-Ethoxy-2-mercaptobenzothiazole, 99%

Catalog No. AAL0444714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0444714 25 g
AAL0444706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0444714 Supplier Thermo Scientific Chemicals Supplier No. L0444714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120-53-6 | C9H9NOS2 | 211.297 g/mol

6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and halogens.

Chemical Identifiers

CAS 120-53-6
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
MDL Number MFCD00005782
InChI Key HOASVNMVYBSLSU-UHFFFAOYSA-N
Synonym 6-ethoxy-2-mercaptobenzothiazole, 6-ethoxybenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-ethoxy, 6-ethoxybenzothiazolethiol, usaf pd-58, 6-ethoxy-2-benzothiazolethiol, 6-ethoxybenzo d thiazole-2 3h-thione, 6-ethoxy-1,3-benzothiazole-2-thiol, 2-mercapto-6-ethoxybenzothiazole, 2-benzothiazolethiol, 6-ethoxy
PubChem CID 719344
IUPAC Name 6-ethoxy-3H-1,3-benzothiazole-2-thione
SMILES CCOC1=CC2=C(C=C1)NC(=S)S2

Specifications

Melting Point 196°C to 201°C
Quantity 25 g
Beilstein 152313
Solubility Information Insoluble in water.
Formula Weight 211.32
Percent Purity 99%
Chemical Name or Material 6-Ethoxy-2-mercaptobenzothiazole
EINECSNumber 204-405-5
RTECSNumber DL6498000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only