missing translation for 'onlineSavingsMsg'
Learn More

6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline 90.0+%, TCI America™

Catalog No. p-7128903
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
E023725G 25 g
E0237500G 500 g
2 options

Catalog No. E023725G

Supplier: TCI America E023725G

Only null left

Chemical Identifiers

CAS 91-53-2
Molecular Formula C14H19NO
Molecular Weight (g/mol) 217.312
MDL Number MFCD00023883
InChI Key DECIPOUIJURFOJ-UHFFFAOYSA-N
Synonym ethoxyquin, 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, ethoxyquine, santoquin, santoquine, niflex, antioxidant ec, santoflex a, santoflex aw, stop-scald
PubChem CID 3293
ChEBI CHEBI:77323
IUPAC Name 6-ethoxy-2,2,4-trimethyl-1H-quinoline
SMILES CCOC1=CC2=C(C=C1)NC(C=C2C)(C)C

Specifications

Boiling Point 161°C
Quantity 25 g
UN Number 3082
Formula Weight 217.31
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline [Antioxidant for rubber]
EINECSNumber (5)-0813
RTECSNumber VB8225000
TSCA Yes