missing translation for 'onlineSavingsMsg'
Learn More

6-Dodecyne 98.0+%, TCI America™

Catalog No. D11925ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11925ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11925ML Supplier TCI America Supplier No. D11925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6975-99-1
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00041658
InChI Key IHPFQAOOSAGSPN-UHFFFAOYSA-N
Synonym 6-dodecyne, acmc-209oav
PubChem CID 138890
IUPAC Name dodec-6-yne
SMILES CCCCCC#CCCCCC

Specifications

Color Yellow
Boiling Point 209°C
Quantity 5 mL
Formula Weight 166.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Dodecyne