missing translation for 'onlineSavingsMsg'
Learn More

6-Dodecyne, 98%

Catalog No. AAL1126206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1126206 5 g
AAL1126214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1126206 Supplier Thermo Scientific Chemicals Supplier No. L1126206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6975-99-1 | C12H22 | 166.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6975-99-1
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00041658
InChI Key IHPFQAOOSAGSPN-UHFFFAOYSA-N
Synonym 6-dodecyne, acmc-209oav
PubChem CID 138890
IUPAC Name dodec-6-yne
SMILES CCCCCC#CCCCCC

Specifications

Density 0.79
Boiling Point 99°C to 100°C (4 mmHg)
Refractive Index 1.439
Linear Formula CH3(CH2)4C≡C(CH2)4CH3
Quantity 5 g
Beilstein 1737397
Formula Weight 166.31
Percent Purity 98%
Chemical Name or Material 6-Dodecyne
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only