missing translation for 'onlineSavingsMsg'
Learn More

6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol 98.0+%, TCI America™

Catalog No. p-7127646
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D326525G 25 g
D3265100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D326525G Supplier TCI America Supplier No. D326525G

Chemical Identifiers

CAS 29529-99-5
Molecular Formula C11H20N4S2
Molecular Weight (g/mol) 272.429
MDL Number MFCD00272594
InChI Key IXDGHAZCSMVIFX-UHFFFAOYSA-N
Synonym 2-(Dibutylamino)-4,6-dimercapto-1,3,5-triazine
PubChem CID 3035346
IUPAC Name 6-(dibutylamino)-1H-1,3,5-triazine-2,4-dithione
SMILES CCCCN(CCCC)C1=NC(=S)NC(=S)N1

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 25 g
Formula Weight 272.43
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol
EINECSNumber (5)-0996
TSCA Yes