Learn More
6-Cyanonicotinic acid, 97%
CAS: 70165-31-0 | C7H4N2O2 | 148.121 g/mol
Supplier: Thermo Scientific Chemicals L17127MD
Description
6-Cyanonicotinic acid, is used in medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Cyanonicotinic acid, is used in medicine
Solubility
Soluble in alcohol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 6-Cyanonicotinic acid | |
| 189°C to 194°C | |
| MFCD01318135 | |
| 4384027 | |
| Soluble in alcohol. | |
| C1=CC(=NC=C1C(=O)O)C#N | |
| 148.121 | |
| 148.12 |
| 70165-31-0 | |
| C7H4N2O2 | |
| 250 mg | |
| 6-cyanonicotinic acid, 2-cyano-5-carboxypyridine, 6-cyanonicotinicacid, 3-pyridinecarboxylic acid, 6-cyano, 5-carboxypicolinonitrile, 3-pyridinecarboxylicacid, 6-cyano, 6-cyano-3-pyridinecarboxylic acid, 6-cyano-nicotinic acid, pubchem16844, 6-cyanonicotinic acid, | |
| WMHSQCDPPJRWIL-UHFFFAOYSA-N | |
| 6-cyanopyridine-3-carboxylic acid | |
| 2761108 | |
| 97% |
Chemical Identifiers
| 70165-31-0 | |
| 148.121 | |
| WMHSQCDPPJRWIL-UHFFFAOYSA-N | |
| 2761108 | |
| C1=CC(=NC=C1C(=O)O)C#N |
| C7H4N2O2 | |
| MFCD01318135 | |
| 6-cyanonicotinic acid, 2-cyano-5-carboxypyridine, 6-cyanonicotinicacid, 3-pyridinecarboxylic acid, 6-cyano, 5-carboxypicolinonitrile, 3-pyridinecarboxylicacid, 6-cyano, 6-cyano-3-pyridinecarboxylic acid, 6-cyano-nicotinic acid, pubchem16844, 6-cyanonicotinic acid, | |
| 6-cyanopyridine-3-carboxylic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only