missing translation for 'onlineSavingsMsg'
Learn More

6-Cyanoindole 98.0+%, TCI America™

Catalog No. p-7125277
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20845G 5 g
C208425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20845G Supplier TCI America Supplier No. C20845G
Only null left

Chemical Identifiers

CAS 15861-36-6
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00016732
InChI Key SZSZDBFJCQKTRG-UHFFFAOYSA-N
Synonym 6-cyanoindole, 6-indolecarbonitrile, indole-6-carbonitrile, 6-cyano-1h-indole, indole-6-cyano, 6-cyano-indole, 6-indole carbonitrile, pubchem7239, cyanoindole-6, 1h-indol-6-yl cyanide
PubChem CID 85146
IUPAC Name 1H-indole-6-carbonitrile
SMILES C1=CC(=CC2=C1C=CN2)C#N

Specifications

Melting Point 130°C
Quantity 5 g
UN Number 3439
Formula Weight 142.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Cyanoindole