missing translation for 'onlineSavingsMsg'
Learn More

6-Cyanoimidazo[1,2-a]pyridine, 95%

Catalog No. AAH3585406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3585406 5 g
AAH3585403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3585406 Supplier Thermo Scientific Chemicals Supplier No. H3585406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106850-34-4 | C8H5N3 | 143.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106850-34-4
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.149
MDL Number MFCD06659603
InChI Key LRJOKNYELSECDZ-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine, 4-hydroimidazo 1,2-a pyridine-6-carbonitrile, 6-cyano-imidazo 1,2-a pyridine, acmc-1bu34, 6-imidazo 1,2-a pyridinecarbonitrile, imidazo 1,2-, a pyridine-6-carbonitrile, imidazo 1,2-alpha pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine-6-carbonitrile
PubChem CID 2764471
IUPAC Name imidazo[1,2-a]pyridine-6-carbonitrile
SMILES C1=CC2=NC=CN2C=C1C#N

Specifications

Melting Point 161°C to 163°C
Quantity 5 g
Formula Weight 148.15
Percent Purity 95%
Chemical Name or Material 6-Cyanoimidazo[1,2-a]pyridine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only