missing translation for 'onlineSavingsMsg'
Learn More

6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one 95.0+%, TCI America™

Catalog No. c2404200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2404200MG 200 mg
C24041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2404200MG Supplier TCI America Supplier No. C2404200MG
Only null left

Chemical Identifiers

CAS 128119-95-9
Molecular Formula C21H17N3O2
Molecular Weight (g/mol) 343.386
InChI Key DOIVGWIGBQTTHP-UHFFFAOYSA-N
PubChem CID 14512392
IUPAC Name 9-(diethylamino)-5-oxobenzo[a]phenoxazine-6-carbonitrile
SMILES CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=O)C(=C3O2)C#N

Specifications

Quantity 200 mg
Formula Weight 343.39
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one