missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chlorouracil 98.0+%, TCI America™
$37.29 - $143.54
Chemical Identifiers
| CAS | 4270-27-3 |
|---|---|
| Molecular Formula | C4H3ClN2O2 |
| Molecular Weight (g/mol) | 146.53 |
| MDL Number | MFCD00014595 |
| InChI Key | PKUFNWPSFCOSLU-UHFFFAOYSA-N |
| Synonym | 6-chlorouracil, 6-chloropyrimidine-2,4 1h,3h-dione, 4-chlorouracil, 6-chloropyrimidine-2,4-diol, 6-chloro-2,4 1h,3h-pyrimidinedione, 6-chloro-2,4-dihydroxypyrimidine, 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-chloro-2,4-pyrimidinediol, 6-chloro-1,3-dihydropyrimidine-2,4-dione, 4-chloro-2,6-dihydroxypyrimidine |
| PubChem CID | 73272 |
| IUPAC Name | 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | ClC1=CC(=O)NC(=O)N1 |
Chemical Identifiers
| 4270-27-3 | |
| 146.53 | |
| PKUFNWPSFCOSLU-UHFFFAOYSA-N | |
| 73272 | |
| ClC1=CC(=O)NC(=O)N1 |
| C4H3ClN2O2 | |
| MFCD00014595 | |
| 6-chlorouracil, 6-chloropyrimidine-2,4 1h,3h-dione, 4-chlorouracil, 6-chloropyrimidine-2,4-diol, 6-chloro-2,4 1h,3h-pyrimidinedione, 6-chloro-2,4-dihydroxypyrimidine, 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-chloro-2,4-pyrimidinediol, 6-chloro-1,3-dihydropyrimidine-2,4-dione, 4-chloro-2,6-dihydroxypyrimidine | |
| 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione |
Specifications
| 4270-27-3 | |
| White-Yellow | |
| MFCD00014595 | |
| 6-chlorouracil, 6-chloropyrimidine-2,4 1h,3h-dione, 4-chlorouracil, 6-chloropyrimidine-2,4-diol, 6-chloro-2,4 1h,3h-pyrimidinedione, 6-chloro-2,4-dihydroxypyrimidine, 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-chloro-2,4-pyrimidinediol, 6-chloro-1,3-dihydropyrimidine-2,4-dione, 4-chloro-2,6-dihydroxypyrimidine | |
| ClC1=CC(=O)NC(=O)N1 | |
| 146.53 | |
| 146.53 | |
| Crystalline Powder |
| 295°C | |
| C4H3ClN2O2 | |
| 5 g | |
| PKUFNWPSFCOSLU-UHFFFAOYSA-N | |
| 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 73272 | |
| ≥98.0% (T) | |
| 6-Chlorouracil |
Safety and Handling
TSCA : No