missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-3-boronic acid, 96%

Catalog No. p-7058545
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2038803 1 g
AAL2038806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2038803 Supplier Thermo Scientific Chemicals Supplier No. L2038803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 444120-91-6 | C5H5BClNO2 | 157.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 444120-91-6
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD03094998
InChI Key WPAPNCXMYWRTTL-UHFFFAOYSA-N
Synonym 2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
PubChem CID 3845034
IUPAC Name (6-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 154°C to 157°C
Quantity 1 g
Beilstein 9119810
Formula Weight 157.37
Percent Purity 96%
Chemical Name or Material 6-Chloropyridine-3-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only