missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-2-carboxaldehyde, 97%

CAS: 54087-03-5 | C6H4ClNO | 141.554 g/mol

Supplier:  Thermo Scientific Chemicals H6412906

 View more versions of this product

Catalog No. AAH6412906

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Chloropyridine-2-carboxaldehyde
69°C to 70°C
C6H4ClNO
5 g
6-chloropicolinaldehyde, 6-chloro-2-pyridinecarbaldehyde, 6-chloro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-chloro, 2-chloro-6-formylpyridine, 6-chloro-2-formylpyridine, 2-chloropyridine-6-carboxaldehyde, 6-chloro-2-pyridinecarboxaldehyde, pubchem13323, chloropyridinecarbaldehyde
C1=CC(=NC(=C1)Cl)C=O
141.554
141.55
Powder
54087-03-5
White
MFCD09832941
1446583
XTRLIKXVRGWTKW-UHFFFAOYSA-N
6-chloropyridine-2-carbaldehyde
10796848
97%

Chemical Identifiers

54087-03-5
141.554
XTRLIKXVRGWTKW-UHFFFAOYSA-N
10796848
C1=CC(=NC(=C1)Cl)C=O
C6H4ClNO
MFCD09832941
6-chloropicolinaldehyde, 6-chloro-2-pyridinecarbaldehyde, 6-chloro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-chloro, 2-chloro-6-formylpyridine, 6-chloro-2-formylpyridine, 2-chloropyridine-6-carboxaldehyde, 6-chloro-2-pyridinecarboxaldehyde, pubchem13323, chloropyridinecarbaldehyde
6-chloropyridine-2-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only