missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-2-carboxaldehyde, 97%

Catalog No. AAH64129MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64129MD 250 mg
AAH6412903 1 g
AAH6412906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64129MD Supplier Thermo Scientific Chemicals Supplier No. H64129MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54087-03-5 | C6H4ClNO | 141.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54087-03-5
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD09832941
InChI Key XTRLIKXVRGWTKW-UHFFFAOYSA-N
Synonym 6-chloropicolinaldehyde, 6-chloro-2-pyridinecarbaldehyde, 6-chloro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-chloro, 2-chloro-6-formylpyridine, 6-chloro-2-formylpyridine, 2-chloropyridine-6-carboxaldehyde, 6-chloro-2-pyridinecarboxaldehyde, pubchem13323, chloropyridinecarbaldehyde
PubChem CID 10796848
IUPAC Name 6-chloropyridine-2-carbaldehyde
SMILES C1=CC(=NC(=C1)Cl)C=O

Specifications

Melting Point 69°C to 70°C
Color White
Quantity 250 mg
Beilstein 1446583
Formula Weight 141.55
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Chloropyridine-2-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only