missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-2-boronic acid pinacol ester, tech. 90%, Thermo Scientific Chemicals

Catalog No. p-7029906
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2742603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2742603 Supplier Thermo Scientific Chemicals Supplier No. H2742603
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 652148-92-0
Molecular Formula C11H15BClNO2
Molecular Weight (g/mol) 239.51
MDL Number MFCD06798259
InChI Key CROJXBLQCOIOBI-UHFFFAOYSA-N
Synonym 6-chloropyridine-2-boronic acid pinacol ester, 2-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-chloropyridin-2-ylboronic acid pinacol ester, 6-chloro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, pyridine, 2-chloro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem21016, 6-chloropyridin-2-boronic acid pinacol ester, 6-chloropyridin-2-yl boronic acid pinacol ester
PubChem CID 17750491
IUPAC Name 2-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC(Cl)=N1

Specifications

Melting Point 107°C to 110°C
Quantity 1 g
Formula Weight 239.51
Percent Purity 90%
Grade Technical
Chemical Name or Material 6-Chloropyridine-2-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only