missing translation for 'onlineSavingsMsg'
Learn More

6-chloropyridazin-3-amine, Thermo Scientific™, 97%, Thermo Scientific™

Catalog No. p-2716144
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB05877EA 10 g
BTB05877EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB05877EA Supplier Thermo Scientific Chemicals Supplier No. BTB05877EA
Only null left

Chemical Identifiers

CAS 5469-69-2
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD00051506
InChI Key DTXVKPOKPFWSFF-UHFFFAOYSA-N
Synonym 3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
PubChem CID 21643
IUPAC Name 6-chloropyridazin-3-amine
SMILES NC1=CC=C(Cl)N=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 129.55
Percent Purity 97%
Chemical Name or Material 6-Chloropyridazin-3-amine
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.