missing translation for 'onlineSavingsMsg'
Learn More

6-chloropurine, 99+%

CAS: 87-42-3 | C5H3ClN4 | 154.56 g/mol

$152.61 - $152.61

Chemical Identifiers

CAS 87-42-3
Molecular Formula C5H3ClN4
Molecular Weight (g/mol) 154.56
MDL Number MFCD00075825
InChI Key ZKBQDFAWXLTYKS-UHFFFAOYSA-N
Synonym 6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
PubChem CID 5359277
IUPAC Name 6-chloro-7H-purine
SMILES C1=NC2=C(N1)C(=NC=N2)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC171280050
SDS View Documents
Thermo Scientific Chemicals
171280050
5 g Glass bottle
Each for $152.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87-42-3
154.56
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
5359277
C1=NC2=C(N1)C(=NC=N2)Cl
C5H3ClN4
MFCD00075825
6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
6-chloro-7H-purine

Specifications

87-42-3
100.0
Orange-Yellow to Yellow
0.5% max. (105°C, 5 hrs)
99+%
C5H3ClN4
5 g
6-chloropurine, 6-chloro-9h-purine, 1h-purine, 6-chloro, 6-chloro-1h-purine, purine, 6-chloro, 6-cp, 6-chlorpurine, 6-chloro purine, 6-chloropurin, chloropurine, 6
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
6-chloro-7H-purine
5359277
99+%
6-chloropurine
99.0
>300°C
(on dry substance) :
Authentic
Glass bottle
MFCD00075825
15, 2163
Solubility in water: 5g/L water
C1=NC2=C(N1)C(=NC=N2)Cl
154.56
154.56
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-745-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only