Learn More
6-Chloropurine 2'-deoxyriboside, 97%
CAS: 4594-45-0 | C10H11ClN4O3 | 270.673 g/mol
Supplier: Thermo Scientific Chemicals J6487103
| Quantity | 1 g |
|---|
Description
6-Chloropurine 2'-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chloropurine 2′-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.
Notes
Light sensitive.
Chemical Identifiers
| C10H11ClN4O3 | |
| MFCD00083282 | |
| 6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine | |
| (2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol |
Specifications
| 6-Chloropurine 2'-deoxyriboside | |
| White | |
| 1 g | |
| Light sensitive | |
| PGEULCIODBNODW-FSDSQADBSA-N | |
| (2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol | |
| 92132234 | |
| 97% |
| 4594-45-0 | |
| C10H11ClN4O3 | |
| MFCD00083282 | |
| 6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine | |
| C1C(C(OC1N2C=NC3=C2N=CN=C3Cl)CO)O | |
| 270.673 | |
| 270.67 | |
| Powder |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Keep cold