missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine 2'-deoxyriboside, 97%

Catalog No. AAJ64871MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64871MD 250 mg
AAJ6487103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64871MD Supplier Thermo Scientific Chemicals Supplier No. J64871MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4594-45-0 | C10H11ClN4O3 | 270.673 g/mol

6-Chloropurine 2'-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloropurine 2′-deoxyriboside is used in the synthesis of 9-(2,3-dideoxy-2-fluoro-beta-D-threo-pentofuranosyl)adenine.

Notes
Light sensitive.

Chemical Identifiers

CAS 4594-45-0
Molecular Formula C10H11ClN4O3
Molecular Weight (g/mol) 270.673
MDL Number MFCD00083282
InChI Key PGEULCIODBNODW-FSDSQADBSA-N
Synonym 6-chloro-9-2'-deoxy-b-d-ribofuranosyl purine
PubChem CID 92132234
IUPAC Name (2R,3R,5R)-5-(6-chloropurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3Cl)CO)O

Specifications

Color White
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 270.67
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Chloropurine 2'-deoxyriboside
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only