missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropiperonyl alcohol, 98%

Catalog No. p-7044348
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0597206 5 g
AAL0597214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0597206 Supplier Thermo Scientific Chemicals Supplier No. L0597206
Only null left

CAS: 2591-25-5 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2591-25-5
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00051727
InChI Key ADHYYNHCXPPVHQ-UHFFFAOYSA-N
Synonym 6-chloropiperonyl alcohol, 6-chloro-2h-1,3-benzodioxol-5-yl methanol, 6-chlorobenzo d 1,3 dioxol-5-yl methanol, 6-chloro-1,3-benzodioxol-5-yl methanol, 2-chlor-4,5-methylendioxy-benzylalkohol, 6-chloro-3,4-methylenedioxybenzyl alcohol, 6-chloro-2h-benzo d 1,3-dioxolen-5-yl methan-1-ol
PubChem CID 7015319
IUPAC Name (6-chloro-1,3-benzodioxol-5-yl)methanol
SMILES C1OC2=C(O1)C=C(C(=C2)CO)Cl

Specifications

Melting Point 68°C to 72°C
Quantity 5 g
Beilstein 150801
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 6-Chloropiperonyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only