missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chlorooxindole 98.0+%, TCI America™
$46.20 - $138.61
Chemical Identifiers
| CAS | 56341-37-8 |
|---|---|
| Molecular Formula | C8H6ClNO |
| Molecular Weight (g/mol) | 167.592 |
| InChI Key | CENVPIZOTHULGJ-UHFFFAOYSA-N |
| Synonym | 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one |
| PubChem CID | 736344 |
| IUPAC Name | 6-chloro-1,3-dihydroindol-2-one |
| SMILES | C1C2=C(C=C(C=C2)Cl)NC1=O |
Chemical Identifiers
| 56341-37-8 | |
| 167.592 | |
| 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one | |
| 6-chloro-1,3-dihydroindol-2-one |
| C8H6ClNO | |
| CENVPIZOTHULGJ-UHFFFAOYSA-N | |
| 736344 | |
| C1C2=C(C=C(C=C2)Cl)NC1=O |
Specifications
| 56341-37-8 | |
| C8H6ClNO | |
| 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one | |
| C1C2=C(C=C(C=C2)Cl)NC1=O | |
| 167.592 | |
| 167.59 | |
| Crystalline Powder |
| 197°C | |
| 5 g | |
| CENVPIZOTHULGJ-UHFFFAOYSA-N | |
| 6-chloro-1,3-dihydroindol-2-one | |
| 736344 | |
| ≥98.0% (GC) | |
| 6-Chlorooxindole |
Safety and Handling
TSCA : No