missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorooxindole 98.0+%, TCI America™

Catalog No. C20245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20245G 5 g
C202425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20245G Supplier TCI America Supplier No. C20245G

Chemical Identifiers

CAS 56341-37-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
InChI Key CENVPIZOTHULGJ-UHFFFAOYSA-N
Synonym 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
PubChem CID 736344
IUPAC Name 6-chloro-1,3-dihydroindol-2-one
SMILES C1C2=C(C=C(C=C2)Cl)NC1=O

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 167.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chlorooxindole