missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorooxindole, 98%

CAS: 56341-37-8 | C8H6ClNO | 167.592 g/mol

Supplier:  Thermo Scientific Chemicals A1967414

 View more versions of this product

Catalog No. AAA1967414


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-Chlorooxindole
195°C to 199°C
MFCD00209962
6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
C1C2=C(C=C(C=C2)Cl)NC1=O
167.592
167.6
56341-37-8
C8H6ClNO
25 g
CENVPIZOTHULGJ-UHFFFAOYSA-N
6-chloro-1,3-dihydroindol-2-one
736344
98%

Chemical Identifiers

56341-37-8
167.592
CENVPIZOTHULGJ-UHFFFAOYSA-N
736344
C1C2=C(C=C(C=C2)Cl)NC1=O
C8H6ClNO
MFCD00209962
6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
6-chloro-1,3-dihydroindol-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only