missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorooxindole, 98%

Catalog No. AAA1967422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1967422 100 g
AAA1967406 5 g
AAA1967414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1967422 Supplier Thermo Scientific Chemicals Supplier No. A1967422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56341-37-8 | C8H6ClNO | 167.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56341-37-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00209962
InChI Key CENVPIZOTHULGJ-UHFFFAOYSA-N
Synonym 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
PubChem CID 736344
IUPAC Name 6-chloro-1,3-dihydroindol-2-one
SMILES C1C2=C(C=C(C=C2)Cl)NC1=O

Specifications

Melting Point 195°C to 199°C
Quantity 100 g
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 6-Chlorooxindole
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only