missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinic Acid 98.0+%, TCI America™

Catalog No. p-7124800
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
C111625G 25 g
C1116100G 100 g
2 options

Catalog No. C111625G

Supplier: TCI America C111625G

Only null left

Chemical Identifiers

CAS 5326-23-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00006241
InChI Key UAWMVMPAYRWUFX-UHFFFAOYSA-N
Synonym 6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid
PubChem CID 79222
IUPAC Name 6-chloropyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 157.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloronicotinic Acid