Learn More
6-Chloronicotinic acid, 99%
CAS: 5326-23-8 | C6H4ClNO2 | 157.553 g/mol
Supplier: Thermo Scientific Chemicals A1562014
Description
6-Chloropyridine-3-carboxylic acid (6-chloronicotinic acid/6-CNA) has been used to study its photolytic and photocatalytic degradation. The product has been used as a media component during the isolation of 6-CNA degrading bacterial strain from imidacloprid-exposed soil samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chloropyridine-3-carboxylic acid (6-chloronicotinic acid/6-CNA) has been used to study its photolytic and photocatalytic degradation. The product has been used as a media component during the isolation of 6-CNA degrading bacterial strain from imidacloprid-exposed soil samples.
Solubility
Soluble in water (2 mg/ml at 20°C), ethanol, methanol, DMSO, and slightly soluble in chloroform.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 6-Chloronicotinic acid | |
| ∼192°C (decomposition) | |
| MFCD00006241 | |
| 115993 | |
| Soluble in water (2mg/ml at 20°C),ethanol,methanol,DMSO,and slightly soluble in chloroform. | |
| C1=CC(=NC=C1C(=O)O)Cl | |
| 157.553 | |
| 157.56 |
| 5326-23-8 | |
| C6H4ClNO2 | |
| 25 g | |
| 6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid | |
| UAWMVMPAYRWUFX-UHFFFAOYSA-N | |
| 6-chloropyridine-3-carboxylic acid | |
| 79222 | |
| 99% |
Chemical Identifiers
| 5326-23-8 | |
| 157.553 | |
| UAWMVMPAYRWUFX-UHFFFAOYSA-N | |
| 79222 | |
| C1=CC(=NC=C1C(=O)O)Cl |
| C6H4ClNO2 | |
| MFCD00006241 | |
| 6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid | |
| 6-chloropyridine-3-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-201-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only