missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinic acid, 99%

Catalog No. AC168691000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168691000 100 g Glass bottle
AC168690050 5 g Glass bottle
AC168690250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC168691000 Supplier Thermo Scientific Chemicals Supplier No. 168691000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5326-23-8 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5326-23-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD00006241
InChI Key UAWMVMPAYRWUFX-UHFFFAOYSA-N
Synonym 6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid
PubChem CID 79222
IUPAC Name 6-chloropyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 186°C to 190°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl(C5H3N)COOH
Quantity 100 g
Beilstein 22, 43
Solubility Information Solubility in water: 2g/L (20°C)
Formula Weight 157.56
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloronicotinic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-201-5
TSCA TSCA

RUO – Research Use Only