missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole 98.0+%, TCI America™

$55.84 - $157.87

Chemical Identifiers

CAS 17422-33-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00005681
InChI Key YTYIMDRWPTUAHP-UHFFFAOYSA-N
Synonym 6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
PubChem CID 87111
ChEBI CHEBI:80918
IUPAC Name 6-chloro-1H-indole
SMILES ClC1=CC=C2C=CNC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C12491G
View Documents
TCI America
C12491G
1 g
Each for $55.84
Only null left
 
C12495G
View Documents
TCI America
C12495G
5 g
Each for $157.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17422-33-2
151.59
YTYIMDRWPTUAHP-UHFFFAOYSA-N
87111
6-chloro-1H-indole
C8H6ClN
MFCD00005681
6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
CHEBI:80918
ClC1=CC=C2C=CNC2=C1

Specifications

17422-33-2
C8H6ClN
1 g
YTYIMDRWPTUAHP-UHFFFAOYSA-N
6-chloro-1H-indole
87111
151.59
Crystalline Powder
89°C
MFCD00005681
6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
ClC1=CC=C2C=CNC2=C1
151.59
CHEBI:80918
≥98.0% (GC)
6-Chloroindole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'