missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole 98.0+%, TCI America™

Catalog No. C12495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12495G 5 g
C12491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12495G Supplier TCI America Supplier No. C12495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17422-33-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00005681
InChI Key YTYIMDRWPTUAHP-UHFFFAOYSA-N
Synonym 6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
PubChem CID 87111
ChEBI CHEBI:80918
IUPAC Name 6-chloro-1H-indole
SMILES ClC1=CC=C2C=CNC2=C1

Specifications

Melting Point 89°C
Quantity 5 g
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloroindole