missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole, 99%

Catalog No. p-7027129
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2365003 1 g
AAB2365006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2365003 Supplier Thermo Scientific Chemicals Supplier No. B2365003
Only null left

CAS: 17422-33-2 | C8H6ClN | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17422-33-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00005681
InChI Key YTYIMDRWPTUAHP-UHFFFAOYSA-N
Synonym 6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
PubChem CID 87111
ChEBI CHEBI:80918
IUPAC Name 6-chloro-1H-indole
SMILES ClC1=CC=C2C=CNC2=C1

Specifications

Melting Point 87°C to 90°C
Quantity 1 g
Beilstein 112345
Formula Weight 151.6
Percent Purity 99%
Chemical Name or Material 6-Chloroindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-449-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only