missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloroindole, 99%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 250mg |
| Solubility Information | Solubility in water: slightly soluble |
| Formula Weight | 151.6 |
| Percent Purity | 99% |
| Chemical Name or Material | 6-Chloroindole, 99% |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
| EINECSNumber | 241-449-4 |
RUO – Research Use Only