missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. AAH6191106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6191106 5 g
AAH6191114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6191106 Supplier Thermo Scientific Chemicals Supplier No. H6191106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 703-82-2
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD06657152
InChI Key CTNIXLBHXMSZKL-UHFFFAOYSA-N
Synonym 6-chloroindole-3-carboxaldehyde, 6-chloro indole-3-carbaldehyde, 6-chloro-3-formylindole, 6-chloro-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 6-chloro, 6-chloro-1h-indole-3-carboxaldehyde, pubchem7693, acmc-209oec, ksc494c8d, 6-chloroindole-3-carbaldehyde
PubChem CID 12614669
IUPAC Name 6-chloro-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C=C1Cl)NC=C2C=O

Specifications

Color Brown
Quantity 5 g
Solubility Information Slightly soluble in water
Formula Weight 179.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Chloroindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only