missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[1,2-a]pyridine 98.0+%, TCI America™

Catalog No. p-7125943
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33701G 1 g
C33705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33701G Supplier TCI America Supplier No. C33701G
Only null left

Chemical Identifiers

CAS 6188-25-6
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD05663807
InChI Key XQEGYCZJSVFGEE-UHFFFAOYSA-N
Synonym 6-chloroimidazo 1,2-a pyridine, 6-chloro-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 6-chloro, chloroimidazoapyridine, 6-chloranylimidazo 1,2-a pyridine, 6-chloroh-imidazo 1,2-a pyridine, 6-chloro-4-hydroimidazo 1,2-a pyridine, 6-chloroimidazo 1 pound not2-a pyridine
PubChem CID 2764328
IUPAC Name 6-chloroimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=CN2C=C1Cl

Specifications

Melting Point 87°C
Boiling Point 132°C
Quantity 1 g
Formula Weight 152.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloroimidazo[1,2-a]pyridine