missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[1,2-a]pyridine, 97%

Catalog No. p-7031951
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3323406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3323406 Supplier Thermo Scientific Chemicals Supplier No. H3323406
Only null left

CAS: 6188-25-6 | C7H5ClN2 | 152.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6188-25-6
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD05663807
InChI Key XQEGYCZJSVFGEE-UHFFFAOYSA-N
Synonym 6-chloroimidazo 1,2-a pyridine, 6-chloro-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 6-chloro, chloroimidazoapyridine, 6-chloranylimidazo 1,2-a pyridine, 6-chloroh-imidazo 1,2-a pyridine, 6-chloro-4-hydroimidazo 1,2-a pyridine, 6-chloroimidazo 1 pound not2-a pyridine
PubChem CID 2764328
IUPAC Name 6-chloroimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=CN2C=C1Cl

Specifications

Melting Point 80°C to 82°C
Quantity 5 g
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 6-Chloroimidazo[1,2-a]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only