missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%

CAS: 138642-97-4 | C8H5ClN2O2 | 196.59 g/mol

Supplier:  Thermo Scientific Chemicals H3556703

 Afficher plus de versions de ce produit

Numéro de catalogue. AAH3556703


Il en reste null
Explore available promotions

Description

Description

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate
C8H5ClN2O2
1 g
Slightly soluble in water.
OC(=O)C1=CN=C2C=CC(Cl)=CN12
196.59
196.59 (Anhydrous)
138642-97-4
MFCD07021509
6-chloroimidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-chloro
LXRDUEDQFSBROM-UHFFFAOYSA-N
6-chloroimidazo[1,2-a]pyridine-3-carboxylic acid
16413639
95%

Chemical Identifiers

138642-97-4
196.59
LXRDUEDQFSBROM-UHFFFAOYSA-N
16413639
OC(=O)C1=CN=C2C=CC(Cl)=CN12
C8H5ClN2O2
MFCD07021509
6-chloroimidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-chloro
6-chloroimidazo[1,2-a]pyridine-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only