missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorochromone 98.0+%, TCI America™

Catalog No. C30395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30395G 5 g
C303925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30395G Supplier TCI America Supplier No. C30395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33533-99-2
Molecular Formula C9H5ClO2
Molecular Weight (g/mol) 180.587
MDL Number MFCD00191904
InChI Key VFZQATFTQAZCMO-UHFFFAOYSA-N
PubChem CID 601989
IUPAC Name 6-chlorochromen-4-one
SMILES C1=CC2=C(C=C1Cl)C(=O)C=CO2

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 180.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chlorochromone