Learn More
6-Chlorobenzimidazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals
$312.47 - $312.47
Chemical Identifiers
| CAS | 39811-14-8 |
|---|---|
| Molecular Formula | C8H5ClN2O2 |
| Molecular Weight (g/mol) | 196.59 |
| MDL Number | MFCD06739053 |
| InChI Key | NZIHMSYSZRFUQJ-UHFFFAOYSA-N |
| Synonym | 6-chloro-1h-benzoimidazole-2-carboxylic acid, 6-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzimidazole-2-carboxylic acid, 6-chlorobenzimidazole-2-carboxylic acid, 5-chloro-1h-benzoimidazole-2-carboxylic acid, 1h-benzimidazole-2-carboxylic acid, 6-chloro, 5-chloro-1h-1,3-benzodiazole-2-carboxylic acid, 5-chloro-3h-1,3-benzodiazole-2-carboxylic acid, 6-chloro-1h-1,3-benzodiazole-2-carboxylic acid |
| PubChem CID | 10512110 |
| IUPAC Name | 6-chloro-1H-benzimidazole-2-carboxylic acid |
| SMILES | C1=CC2=C(C=C1Cl)NC(=N2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5864803
|
Thermo Scientific Chemicals
H5864803 |
1 g |
Each for $312.47
|
|
|||||
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
This products exists as two tautomers as there is a rapid exchange of the proton between the two heterocyclic nitrogens. This renders the 5- and 6- positions on the ring chemically equivalent.
Chemical Identifiers
| 39811-14-8 | |
| 196.59 | |
| NZIHMSYSZRFUQJ-UHFFFAOYSA-N | |
| 10512110 | |
| C1=CC2=C(C=C1Cl)NC(=N2)C(=O)O |
| C8H5ClN2O2 | |
| MFCD06739053 | |
| 6-chloro-1h-benzoimidazole-2-carboxylic acid, 6-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzimidazole-2-carboxylic acid, 6-chlorobenzimidazole-2-carboxylic acid, 5-chloro-1h-benzoimidazole-2-carboxylic acid, 1h-benzimidazole-2-carboxylic acid, 6-chloro, 5-chloro-1h-1,3-benzodiazole-2-carboxylic acid, 5-chloro-3h-1,3-benzodiazole-2-carboxylic acid, 6-chloro-1h-1,3-benzodiazole-2-carboxylic acid | |
| 6-chloro-1H-benzimidazole-2-carboxylic acid |
Specifications
| 39811-14-8 | |
| MFCD06739053 | |
| 6-chloro-1h-benzoimidazole-2-carboxylic acid, 6-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzimidazole-2-carboxylic acid, 6-chlorobenzimidazole-2-carboxylic acid, 5-chloro-1h-benzoimidazole-2-carboxylic acid, 1h-benzimidazole-2-carboxylic acid, 6-chloro, 5-chloro-1h-1,3-benzodiazole-2-carboxylic acid, 5-chloro-3h-1,3-benzodiazole-2-carboxylic acid, 6-chloro-1h-1,3-benzodiazole-2-carboxylic acid | |
| NZIHMSYSZRFUQJ-UHFFFAOYSA-N | |
| 6-chloro-1H-benzimidazole-2-carboxylic acid | |
| 10512110 | |
| 97% |
| C8H5ClN2O2 | |
| 1 g | |
| Slightly soluble in water. | |
| C1=CC2=C(C=C1Cl)NC(=N2)C(=O)O | |
| 196.59 | |
| 196.59 | |
| 6-Chlorobenzimidazole-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only