missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorobenzimidazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH5864803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5864803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5864803 Supplier Thermo Scientific Chemicals Supplier No. H5864803

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
This products exists as two tautomers as there is a rapid exchange of the proton between the two heterocyclic nitrogens. This renders the 5- and 6- positions on the ring chemically equivalent.

Chemical Identifiers

CAS 39811-14-8
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD06739053
InChI Key NZIHMSYSZRFUQJ-UHFFFAOYSA-N
Synonym 6-chloro-1h-benzoimidazole-2-carboxylic acid, 6-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzimidazole-2-carboxylic acid, 6-chlorobenzimidazole-2-carboxylic acid, 5-chloro-1h-benzoimidazole-2-carboxylic acid, 1h-benzimidazole-2-carboxylic acid, 6-chloro, 5-chloro-1h-1,3-benzodiazole-2-carboxylic acid, 5-chloro-3h-1,3-benzodiazole-2-carboxylic acid, 6-chloro-1h-1,3-benzodiazole-2-carboxylic acid
PubChem CID 10512110
IUPAC Name 6-chloro-1H-benzimidazole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Cl)NC(=N2)C(=O)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 196.59
Percent Purity 97%
Chemical Name or Material 6-Chlorobenzimidazole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only