missing translation for 'onlineSavingsMsg'
Learn More

6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one 98.0+%, TCI America™

Catalog No. p-7125827
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30975G 5 g
C309725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30975G Supplier TCI America Supplier No. C30975G

Chemical Identifiers

CAS 26518-76-3
Molecular Formula C10H8ClNO3
Molecular Weight (g/mol) 225.63
MDL Number MFCD01321312
InChI Key MIAHXWVABDHISZ-UHFFFAOYSA-N
PubChem CID 2764458
IUPAC Name 6-(2-chloroacetyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one
SMILES ClCC(=O)C1=CC=C2OCC(=O)NC2=C1

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 5 g
Formula Weight 225.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one